ENA413171438 Enamine

(3-{[methyl(phenyl)amino]methyl}phenyl)boronic acid hydrochloride

  • MDL number MFCD27501011

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155763

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155763
Availability details for
Quantity
  • Availability
  • Add to Cart