ENA413171440 Enamine

(3S)-3-(tert-butoxy)pyrrolidine; oxalic acid

  • MDL number MFCD27501012
  • Cas Number : 1807940-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HOOCCOOH
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155792
Availability details for
Quantity
  • Availability
  • Add to Cart