ENA413171443 Enamine

N-(4-fluorophenyl)azetidin-3-amine dihydrochloride

  • MDL number MFCD27501014
  • Cas Number : 1803590-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-155839

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155839
Availability details for
Quantity
  • Availability
  • Add to Cart