ENA413171444 Enamine

4-(difluoromethyl)piperidin-4-ol hydrochloride

  • MDL number MFCD27920236
  • Cas Number : 1803595-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155850
Availability details for
Quantity
  • Availability
  • Add to Cart