ENA413171453 Enamine

[2-(4-bromophenyl)propan-2-yl](2-methylpropyl)amine hydrochloride

  • MDL number MFCD27920243
  • Cas Number : 1607277-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155963

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155963
Availability details for
Quantity
  • Availability
  • Add to Cart