ENA413171457 Enamine

(3-ethylpiperidin-3-yl)methanol hydrochloride

  • MDL number MFCD27920248
  • Cas Number : 1797816-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155990

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155990
Availability details for
Quantity
  • Availability
  • Add to Cart