ENA413171458 Enamine

ethyl 2-(7-{[(tert-butoxy)carbonyl]amino}-4,5,6,7-tetrahydro-1H-indazol-1-yl)acetate

  • MDL number MFCD27920249
  • Cas Number : 1803582-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155996

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155996
Availability details for
Quantity
  • Availability
  • Add to Cart