ENA413171459 Enamine

methyl 2-amino-3-(1H-imidazol-1-yl)propanoate dihydrochloride

  • MDL number MFCD27920251
  • Cas Number : 1607247-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156010

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156010
Availability details for
Quantity
  • Availability
  • Add to Cart