ENA413171463 Enamine

methyl 2-({[4-(trifluoromethyl)phenyl]methyl}amino)acetate hydrochloride

  • MDL number MFCD27920256
  • Cas Number : 1797551-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156060
Availability details for
Quantity
  • Availability
  • Add to Cart