ENA413171465 Enamine

ethyl 2-amino-3-sulfamoylpropanoate hydrochloride

  • MDL number MFCD27920258
  • Cas Number : 2366201-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156068

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156068
Availability details for
Quantity
  • Availability
  • Add to Cart