ENA413171472 Enamine

(1R,2R)-1-N,1-N-dimethylcyclopentane-1,2-diamine dihydrochloride

  • MDL number MFCD27920266
  • Cas Number : 1807914-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156191

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156191
Availability details for
Quantity
  • Availability
  • Add to Cart