ENA413171473 Enamine

3-(4-bromo-3-methylphenyl)pentanedioic acid

  • MDL number MFCD27920267
  • Cas Number : 1797015-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156201

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156201
Availability details for
Quantity
  • Availability
  • Add to Cart