ENA413171485 Enamine

[(2-aminoethyl)sulfamoyl](2,2,2-trifluoroethyl)amine hydrochloride

  • MDL number MFCD27920279
  • Cas Number : 1797189-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156329

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156329
Availability details for
Quantity
  • Availability
  • Add to Cart