ENA413171488 Enamine

5-bromo-3-methoxy-2-(methylsulfanyl)pyridine

  • MDL number MFCD27920282
  • Cas Number : 1446002-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156383

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156383
Availability details for
Quantity
  • Availability
  • Add to Cart