ENA413171497 Enamine

tert-butyl N-[2-(4-methylbenzoyl)furan-3-yl]carbamate

  • MDL number MFCD27920293
  • Cas Number : 1803605-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156579
Availability details for
Quantity
  • Availability
  • Add to Cart