ENA413171502 Enamine

2-amino-3-(1,5-dimethyl-1H-pyrazol-4-yl)propanoic acid hydrochloride

  • MDL number MFCD27920299
  • Cas Number : 1796882-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156640

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156640
Availability details for
Quantity
  • Availability
  • Add to Cart