ENA413171504 Enamine

methyl N-[4-(fluorosulfonyl)-2-nitrophenyl]carbamate

  • MDL number MFCD27920301
  • Cas Number : 1803600-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156658

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156658
Availability details for
Quantity
  • Availability
  • Add to Cart