ENA413171508 Enamine

6-cyclopentylimidazo[2,1-b][1,3,4]thiadiazole

  • MDL number MFCD27920305
  • Cas Number : 1697870-38-4
  • Molecular Weight : 193.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156705
Availability details for
Quantity
  • Availability
  • Add to Cart