ENA413171510 Enamine

3-(difluoromethoxy)azetidine hydrochloride

  • MDL number MFCD27920307
  • Cas Number : 1619991-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156761
Availability details for
Quantity
  • Availability
  • Add to Cart