ENA413171512 Enamine

2-(3-chlorophenyl)-5-methylmorpholine hydrochloride

  • MDL number MFCD27920310
  • Cas Number : 1803561-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156859

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156859
Availability details for
Quantity
  • Availability
  • Add to Cart