ENA413171516 Enamine

5-(2-aminoethyl)-1,3,4-oxadiazol-2-amine dihydrochloride

  • MDL number MFCD27920314
  • Cas Number : 1797101-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156886

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156886
Availability details for
Quantity
  • Availability
  • Add to Cart