ENA413171518 Enamine

3-methyl-2-[(trimethylsilyl)oxy]but-3-enenitrile

  • MDL number MFCD27920316
  • Cas Number : 60550-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156900
Availability details for
Quantity
  • Availability
  • Add to Cart