ENA413171519 Enamine

5,6-dimethyl-3-(piperazin-1-yl)-1,2,4-triazine hydrochloride

  • MDL number MFCD27920317
  • Cas Number : 1797883-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156906
Availability details for
Quantity
  • Availability
  • Add to Cart