ENA413171522 Enamine

methyl decahydroquinoline-2-carboxylate hydrochloride

  • MDL number MFCD27920320
  • Cas Number : 83240-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156948

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156948
Availability details for
Quantity
  • Availability
  • Add to Cart