ENA413171528 Enamine

methyl 3-(piperazin-1-yl)oxetane-3-carboxylate dihydrochloride

  • MDL number MFCD27920326
  • Cas Number : 1803566-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156978
Availability details for
Quantity
  • Availability
  • Add to Cart