ENA413171534 Enamine

3-(4-amino-5-propyl-1H-pyrazol-1-yl)benzonitrile hydrochloride

  • MDL number MFCD27920333
  • Cas Number : 1797731-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157134

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157134
Availability details for
Quantity
  • Availability
  • Add to Cart