ENA413171539 Enamine

2-(3-amino-1,1-dioxo-1lambda6-thiolan-3-yl)acetic acid hydrochloride

  • MDL number MFCD27920338
  • Cas Number : 1803582-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157154
Availability details for
Quantity
  • Availability
  • Add to Cart