ENA413171542 Enamine

3-(2,2,2-trifluoroethoxy)azetidine hydrochloride

  • MDL number MFCD27920341
  • Cas Number : 1332886-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157259
Availability details for
Quantity
  • Availability
  • Add to Cart