ENA413171545 Enamine

3-(1,1-difluoroethyl)aniline hydrochloride

  • MDL number MFCD27920346
  • Cas Number : 1803593-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157314

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157314
Availability details for
Quantity
  • Availability
  • Add to Cart