ENA413171547 Enamine

4-[(4-aminopiperidin-1-yl)methyl]-2-ethoxyphenol dihydrochloride

  • MDL number MFCD27920348
  • Cas Number : 1797809-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157341

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157341
Availability details for
Quantity
  • Availability
  • Add to Cart