ENA413171550 Enamine

2-[2-(2-methyl-1,3-thiazol-4-yl)ethyl]-2,3-dihydro-1H-isoindole-1,3-dione hydrochloride

  • MDL number MFCD27920351
  • Cas Number : 1803561-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157444

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157444
Availability details for
Quantity
  • Availability
  • Add to Cart