ENA413171556 Enamine

1-amino-3-ethoxy-2,2-dimethylcyclobutane-1-carboxylic acid hydrochloride

  • MDL number MFCD27920357
  • Cas Number : 1796903-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157487

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157487
Availability details for
Quantity
  • Availability
  • Add to Cart