ENA413171558 Enamine

methyl 1-amino-3-ethoxy-2,2-dimethylcyclobutane-1-carboxylate hydrochloride

  • MDL number MFCD27920359
  • Cas Number : 1796887-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157490

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157490
Availability details for
Quantity
  • Availability
  • Add to Cart