ENA413171561 Enamine

methyl 2-bromo-2,3-dimethylbutanoate

  • MDL number MFCD27920362
  • Cas Number : 91524-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157529

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157529
Availability details for
Quantity
  • Availability
  • Add to Cart