ENA413171563 Enamine

1-benzyl-2-phenyl-1H,2H,3H-imidazo[4,5-c]pyridine

  • MDL number MFCD27920364
  • Cas Number : 1803596-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157563

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157563
Availability details for
Quantity
  • Availability
  • Add to Cart