ENA413171564 Enamine

3-(cyclopentylmethyl)pyrrolidine hydrochloride

  • MDL number MFCD27920365
  • Cas Number : 1803597-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157617

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157617
Availability details for
Quantity
  • Availability
  • Add to Cart