ENA413171581 Enamine

5-(propan-2-yl)morpholin-3-one

  • MDL number MFCD18250290
  • Cas Number : 119844-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155453

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155453
Availability details for
Quantity
  • Availability
  • Add to Cart