ENA413171584 Enamine

1-(5-amino-2,3-dihydro-1H-indol-1-yl)-2-methoxyethan-1-one

  • MDL number MFCD13808927
  • Cas Number : 1019498-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155035

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155035
Availability details for
Quantity
  • Availability
  • Add to Cart