ENA413171587 Enamine

5-amino-6-chloropyridine-3-carbonitrile

  • MDL number MFCD18250728
  • Cas Number : 1260672-23-8
  • Molecular Weight : 153.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-amino-6-chloronicotinonitrile
Inchi Code 1S/C6H4ClN3/c7-6-5(9)1-4(2-8)3-10-6/h1,3H,9H2
Inchi Key FAJCKRPVMJRUJX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-155041

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-155041
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart