ENA413171588 Enamine

2-methyl-5-(piperidin-3-yloxy)pyridine

  • MDL number MFCD08688390
  • Cas Number : 946681-30-7
  • Molecular Weight : 192.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155476
Availability details for
Quantity
  • Availability
  • Add to Cart