ENA413171606 Enamine

N-ethyl-1H-1,2,3-triazole-4-carboxamide

  • MDL number MFCD19597205
  • Cas Number : 1342762-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155517
Availability details for
Quantity
  • Availability
  • Add to Cart