ENA413171610 Enamine

N-benzylcyclohexanecarboxamide

  • MDL number MFCD01355913
  • Cas Number : 35665-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155536

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155536
Availability details for
Quantity
  • Availability
  • Add to Cart