ENA413171612 Enamine

1-(2-methyl-6-nitrophenyl)ethan-1-one

  • MDL number MFCD18431418
  • Cas Number : 4127-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155539

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155539
Availability details for
Quantity
  • Availability
  • Add to Cart