ENA413171613 Enamine

3-phenyl-5-(trichloromethyl)-1,2,4-oxadiazole

  • MDL number MFCD01710588
  • Cas Number : 1208-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155541

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155541
Availability details for
Quantity
  • Availability
  • Add to Cart