ENA413171614 Enamine

(3-methoxyphenyl)(4-methylphenyl)methanone

  • MDL number MFCD01311558
  • Cas Number : 82520-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155552

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155552
Availability details for
Quantity
  • Availability
  • Add to Cart