ENA413171625 Enamine

2-(acetyloxy)-2-methylpropanoic acid

  • MDL number MFCD19228148
  • Cas Number : 15805-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-60
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155589
Availability details for
Quantity
  • Availability
  • Add to Cart