ENA413171626 Enamine

5-(phenoxymethyl)-2H-1,2,3,4-tetrazole

  • MDL number MFCD07345675
  • Cas Number : 24896-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155590

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155590
Availability details for
Quantity
  • Availability
  • Add to Cart