ENA413171642 Enamine

5-chloro-2-[(oxolan-3-yl)methoxy]aniline

  • MDL number MFCD14621690
  • Cas Number : 1249299-13-5
  • Molecular Weight : 227.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-2-(tetrahydro-3-furanylmethoxy)aniline
Inchi Code 1S/C11H14ClNO2/c12-9-1-2-11(10(13)5-9)15-7-8-3-4-14-6-8/h1-2,5,8H,3-4,6-7,13H2
Inchi Key WLUQKEUGNJBYRE-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-155652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155652
Availability details for
Quantity
  • Availability
  • Add to Cart