ENA413171648 Enamine

naphthalen-2-yl chloromethanethioate

  • MDL number MFCD02259394
  • Cas Number : 10506-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-72
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-154917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154917
Availability details for
Quantity
  • Availability
  • Add to Cart