ENA413171657 Enamine

1-(5-bromo-2-hydroxy-3-methylphenyl)ethan-1-one

  • MDL number MFCD18294026
  • Cas Number : 72422-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155122
Availability details for
Quantity
  • Availability
  • Add to Cart